Assay Details
Target Gene Details
Entrez Gene ID: | 10602 |
Gene Name: | CDC42 effector protein 3 |
Gene Aliases: |
BORG2, CEP3, UB1 |
Location: |
Chr.2:37641882-37672535 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP3 | NM_001270436.1 | 2 | 3437 | NP_001257365.1 |
| NM_001270437.1 | 2 | 3393 | NP_001257366.1 | |
| NM_001270438.1 | 2 | 3369 | NP_001257367.1 | |
| NM_006449.4 | 2 | 4028 | NP_006440.2 | |
| XM_011532482.2 | 2 | 3834 | XP_011530784.1 | |
| AK055915.1 | 1 | 568 | ||
| BQ432066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833825 | Chr.2:37581026 - 37758774 on Build GRCh38 | Gain |
|
| esv2759043 | Chr.2:37569511 - 37911796 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114822846] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map