Assay Details
Target Gene Details
Entrez Gene ID: | 9947 |
Gene Name: | MAGE family member C1 |
Gene Aliases: |
CT7, CT7.1 |
Location: |
Chr.X:141903856-141909401 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEC1 | NM_005462.4 | NP_005453.2 | ||
| XM_011531418.2 | XP_011529720.1 | |||
| AF056334.1 | AAC24227.1 | |||
| BQ772869.1 |
More Information
Set Membership: |
|

Genomic Map