Assay Details
Target Gene Details
Entrez Gene ID: | 283579 |
Gene Name: | chromosome 14 open reading frame 178 |
Gene Aliases: |
- |
Location: |
Chr.14:77760830-77769742 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C14orf178 | NM_001173978.1 | NP_001167449.1 | ||
| NM_174943.3 | NP_777603.1 | |||
| AK098842.1 | BAC05432.1 | |||
| BC113929.1 | AAI13930.1 |
Target Gene Details
Entrez Gene ID: | 22938 |
Gene Name: | SNW domain containing 1 |
Gene Aliases: |
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP |
Location: |
Chr.14:77717599-77761220 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNW1 | XM_005267413.3 | XP_005267470.1 | ||
| AK316087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759998 | Chr.14:77607837 - 77796229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map