Assay Details
Target Gene Details
Entrez Gene ID: | 8706 |
Gene Name: | beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group) |
Gene Aliases: |
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3 |
Location: |
Chr.3:161083883-161105528 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592128 | Chr.3:160795732 - 161829856 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map