Assay Details
Target Gene Details
Entrez Gene ID: | 29079 |
Gene Name: | mediator complex subunit 4 |
Gene Aliases: |
ARC36, DRIP36, HSPC126, TRAP36, VDRIP |
Location: |
Chr.13:48075728-48095141 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED4 | NM_001270629.1 | 7 | 1241 | NP_001257558.1 |
| NM_014166.3 | 7 | 1347 | NP_054885.1 | |
| XM_017020549.1 | 7 | 2435 | XP_016876038.1 | |
| AF161475.1 | 7 | 1309 | AAF29090.1 | |
| AF300618.1 | 7 | 1284 | AAG22542.1 | |
| AK001934.1 | 7 | 1310 | BAA91987.1 | |
| AK301835.1 | 7 | 1215 | ||
| BC005189.1 | 7 | 1321 | AAH05189.1 | |
| HG518167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832601 | Chr.13:47995817 - 48161803 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map