Assay Details
Target Gene Details
Entrez Gene ID: | 6999 |
Gene Name: | tryptophan 2,3-dioxygenase |
Gene Aliases: |
TDO, TO, TPH2, TRPO |
Location: |
Chr.4:155903693-155920406 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDO2 | NM_005651.3 | NP_005642.1 | ||
| AK289418.1 | ||||
| AK307741.1 | ||||
| BC005355.1 | AAH05355.1 | |||
| BG616664.1 | ||||
| U32989.1 | AAB08514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575777 | Chr.4:155855410 - 156699594 on Build GRCh38 | Gain |
|
| nsv830121 | Chr.4:155850003 - 156002314 on Build GRCh38 | Gain |
|
| nsv1029122 | Chr.4:155773161 - 156031641 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map