Assay Details
Target Gene Details
Entrez Gene ID: | 79290 |
Gene Name: | olfactory receptor family 13 subfamily A member 1 |
Gene Aliases: |
- |
Location: |
Chr.10:45302294-45315608 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR13A1 | NM_001004297.2 | 4 | 1697 | NP_001004297.2 |
| XM_011540147.2 | 3 | 2164 | XP_011538449.1 | |
| XM_017016632.1 | 4 | 5083 | XP_016872121.1 | |
| XM_017016633.1 | 3 | 5225 | XP_016872122.1 | |
| XM_017016634.1 | 3 | 5470 | XP_016872123.1 | |
| AK131555.1 | 4 | 1697 | BAD18689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623082 | Chr.10:45074561 - 45309033 on Build GRCh38 | Gain |
|
| esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain |
|
| esv2750897 | Chr.10:45054697 - 45324688 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map