Assay Details
Target Gene Details
Entrez Gene ID: | 10579 |
Gene Name: | transforming acidic coiled-coil containing protein 2 |
Gene Aliases: |
AZU-1, ECTACC |
Location: |
Chr.10:121989174-122254545 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832009 | Chr.10:122014968 - 122189541 on Build GRCh38 | Loss |
|
| nsv1045815 | Chr.10:121986932 - 122086108 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map