Assay Details
Target Gene Details
Entrez Gene ID: | 5861 |
Gene Name: | RAB1A, member RAS oncogene family |
Gene Aliases: |
RAB1, YPT1 |
Location: |
Chr.2:65086854-65130301 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB1A | NM_004161.4 | NP_004152.1 | ||
| NM_015543.1 | NP_056358.1 | |||
| XM_005264468.1 | XP_005264525.1 | |||
| AF498929.1 | AAM21077.1 | |||
| AK055927.1 | BAB71048.1 | |||
| AK303678.1 | ||||
| AK315495.1 | ||||
| AL050268.1 | CAB43369.1 | |||
| BC000905.2 | AAH00905.1 | |||
| BT019528.1 | AAV38335.1 | |||
| BT019529.1 | AAV38336.1 | |||
| BX571747.1 | CAE11872.1 | |||
| CR533479.1 | CAG38510.1 | |||
| CR536488.1 | CAG38727.1 | |||
| M28209.1 | AAA60240.1 | |||
| X13898.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819395 | Chr.2:65087568 - 65091059 on Build GRCh38 | Loss |
|
| nsv834246 | Chr.2:64981876 - 65176937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map