Assay Details
Target Gene Details
Entrez Gene ID: | 9452 |
Gene Name: | integral membrane protein 2A |
Gene Aliases: |
BRICD2A, E25A |
Location: |
Chr.X:79360384-79367552 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITM2A | NM_001171581.1 | 5 | 1184 | NP_001165052.1 |
| NM_004867.4 | 6 | 1316 | NP_004858.1 | |
| AK223327.1 | 6 | 1095 | BAD97047.1 | |
| AK303238.1 | 5 | 966 | ||
| AY359051.1 | 6 | 1057 | AAQ89410.1 | |
| BC040437.1 | 6 | 1071 | AAH40437.1 | |
| BM561934.1 | 3 | 559 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519126 | Chr.X:79331247 - 79373681 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140404473] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map