Assay Details
Target Gene Details
Entrez Gene ID: | 128239 |
Gene Name: | IQ motif containing GTPase activating protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:156525405-156572604 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQGAP3 | NM_178229.4 | NP_839943.2 | ||
| XM_011509198.2 | XP_011507500.1 | |||
| XM_011509200.2 | XP_011507502.1 | |||
| XM_011509201.2 | XP_011507503.1 | |||
| XM_017000317.1 | XP_016855806.1 | |||
| XM_017000318.1 | XP_016855807.1 | |||
| AB105103.1 | BAC78211.1 | |||
| AK122653.1 | BAC85501.1 | |||
| AY253300.1 | AAP06954.1 | |||
| BC131536.1 |
More Information
Set Membership: |
|

Genomic Map