Assay Details
Target Gene Details
Entrez Gene ID: | 4927 |
Gene Name: | nucleoporin 88 |
Gene Aliases: |
- |
Location: |
Chr.17:5384833-5419739 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP88 | NM_001320653.1 | 15 | 2112 | NP_001307582.1 |
| NM_002532.5 | 15 | 2064 | NP_002523.2 | |
| XM_011523893.2 | 16 | 2162 | XP_011522195.1 | |
| XM_017024694.1 | 16 | 2114 | XP_016880183.1 | |
| AK298158.1 | 16 | 1889 | ||
| AK308142.1 | 15 | 1877 | ||
| BC000335.2 | 15 | 1977 | AAH00335.1 | |
| BF594453.1 | 3 | 455 | ||
| CX870338.1 | 2 | 147 | ||
| Y08612.2 | 15 | 2023 | CAA69904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv819549 | Chr.17:5386112 - 5388850 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map