Assay Details
Target Gene Details
Entrez Gene ID: | 84660 |
Gene Name: | coiled-coil domain containing 62 |
Gene Aliases: |
CT109, ERAP75, TSP-NY |
Location: |
Chr.12:122773621-122827385 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC62 | NM_201435.4 | 9 | 1241 | NP_958843.2 |
| NR_027918.2 | 9 | 1241 | ||
| XM_006719643.1 | 9 | 1149 | XP_006719706.1 | |
| XM_006719644.1 | 9 | 1149 | XP_006719707.1 | |
| XM_011538856.1 | 9 | 1146 | XP_011537158.1 | |
| XM_011538857.1 | 9 | 1149 | XP_011537159.1 | |
| XM_011538859.1 | 6 | 612 | XP_011537161.1 | |
| XM_017020033.1 | 8 | 2027 | XP_016875522.1 | |
| AK097663.1 | 9 | 1165 | ||
| AK124633.1 | 5 | 1565 | BAC85909.1 | |
| AK292466.1 | 9 | 1149 | ||
| AY009105.1 | 5 | 557 | AAG49396.1 | |
| AY254201.1 | 9 | 1222 | AAP13075.1 | |
| BC060796.1 | 9 | 1224 | AAH60796.1 |
More Information
Set Membership: |
|

Genomic Map