Assay Details
Target Gene Details
Entrez Gene ID: | 153527 |
Gene Name: | zinc finger matrin-type 2 |
Gene Aliases: |
Ptg-12, Snu23 |
Location: |
Chr.5:140700360-140706681 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMAT2 | NM_144723.2 | NP_653324.1 | ||
| AK055683.1 | BAB70983.1 | |||
| BC056668.1 | AAH56668.1 | |||
| BP210985.1 | ||||
| BP370587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830508 | Chr.5:140675388 - 140869073 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map