Assay Details
Target Gene Details
Entrez Gene ID: | 535 |
Gene Name: | ATPase H+ transporting V0 subunit a1 |
Gene Aliases: |
ATP6N1, ATP6N1A, Stv1, VPP1, Vph1, a1 |
Location: |
Chr.17:42458844-42522579 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V0A1 | NM_001130020.1 | NP_001123492.1 | ||
| NM_001130021.1 | NP_001123493.1 | |||
| NM_005177.3 | NP_005168.2 | |||
| XM_005257459.1 | XP_005257516.1 | |||
| XM_005257461.1 | XP_005257518.1 | |||
| XM_005257463.2 | XP_005257520.1 | |||
| XM_011524908.1 | XP_011523210.1 | |||
| XM_017024766.1 | XP_016880255.1 | |||
| XM_017024767.1 | XP_016880256.1 | |||
| XM_017024768.1 | XP_016880257.1 | |||
| XM_017024769.1 | XP_016880258.1 | |||
| XM_017024770.1 | XP_016880259.1 | |||
| XM_017024771.1 | XP_016880260.1 | |||
| XM_017024774.1 | XP_016880263.1 | |||
| AK223554.1 | BAD97274.1 | |||
| AK294512.1 | ||||
| AK295682.1 | ||||
| AK299784.1 | ||||
| AK316305.1 | ||||
| BC032398.1 | AAH32398.1 | |||
| BX648978.1 | CAI56709.1 | |||
| CR627443.1 | CAH10528.1 | |||
| L78933.1 | AAL77442.1 | |||
| Z71460.1 | CAA96077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
| nsv575051 | Chr.17:42317580 - 42498404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map