Assay Details
Target Gene Details
Entrez Gene ID: | 162998 |
Gene Name: | olfactory receptor family 7 subfamily D member 2 |
Gene Aliases: |
HTPCRH03, OR19-10, OR19-4 |
Location: |
Chr.19:9185594-9188817 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7D2 | NM_175883.2 | 1 | 1143 | NP_787079.1 |
| AK095468.1 | 1 | 1143 | BAC04554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2411 | Chr.19:9157073 - 9202645 on Build GRCh38 | Deletion |
|
| nsv509720 | Chr.19:9072760 - 9188811 on Build GRCh38 | Insertion |
|
| esv2762024 | Chr.19:8567703 - 9197018 on Build GRCh38 | Gain |
|
| nsv523472 | Chr.19:9180514 - 9340901 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map