Assay Details
Target Gene Details
Entrez Gene ID: | 56985 |
Gene Name: | ADP-ribose/CDP-alcohol diphosphatase, manganese dependent |
Gene Aliases: |
C17orf48, MDS006, NBLA03831 |
Location: |
Chr.17:10697610-10711558 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPRM | NM_020233.4 | 2 | 565 | NP_064618.3 |
| XM_005256738.2 | 2 | 565 | XP_005256795.1 | |
| AB073393.1 | 2 | 536 | BAE45723.1 | |
| AF168715.1 | 1 | 529 | AAF87317.1 | |
| AK292629.1 | 2 | 531 | ||
| DA305828.1 | ||||
| KF880964.1 | 1 | 490 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
|
| nsv1063900 | Chr.17:10662646 - 10729182 on Build GRCh38 | Gain |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map