Assay Details
Target Gene Details
Entrez Gene ID: | 219874 |
Gene Name: | olfactory receptor family 6 subfamily T member 1 |
Gene Aliases: |
OR11-277 |
Location: |
Chr.11:123941474-123943844 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6T1 | NM_001005187.1 | NP_001005187.1 | ||
| XM_017017325.1 | XP_016872814.1 | |||
| BC136906.1 | 1 | 74 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv82n21 | Chr.11:123901052 - 124289279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map