Assay Details
Target Gene Details
Entrez Gene ID: | 9936 |
Gene Name: | CD302 molecule |
Gene Aliases: |
BIMLEC, CLEC13A, DCL-1, DCL1 |
Location: |
Chr.2:159768628-159798255 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD302 | NM_001198763.1 | NP_001185692.1 | ||
| NM_001198764.1 | 3 | 322 | NP_001185693.1 | |
| NM_014880.4 | 4 | 433 | NP_055695.2 | |
| AF499000.1 | 4 | 387 | AAN77050.1 | |
| AK291279.1 | 4 | 386 | ||
| AK304422.1 | ||||
| AY314007.1 | 4 | 420 | AAP79900.1 | |
| BC020646.1 | 3 | 269 | AAH20646.1 | |
| D14664.1 | 4 | 374 | BAA03498.1 | |
| DB217404.1 | 4 | 433 | ||
| DB460994.1 | 3 | 289 |
Target Gene Details
Entrez Gene ID: | 100526664 |
Gene Name: | LY75-CD302 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:159768628-159904756 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LY75-CD302 | NM_001198759.1 | 37 | 5373 | NP_001185688.1 |
| NM_001198760.1 | 36 | 5205 | NP_001185689.1 | |
| AY184222.1 | 37 | 5299 | AAN85434.1 | |
| AY314006.1 | 36 | 5131 | AAP79899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834436 | Chr.2:159765830 - 159941264 on Build GRCh38 | Loss |
|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
| nsv470497 | Chr.2:159694632 - 159836250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map