Assay Details
Target Gene Details
Entrez Gene ID: | 4760 |
Gene Name: | neuronal differentiation 1 |
Gene Aliases: |
BETA2, BHF-1, MODY6, NEUROD, bHLHa3 |
Location: |
Chr.2:181676106-181680665 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEUROD1 | NM_002500.4 | 2 | 2562 | NP_002491.2 |
| AB593068.1 | 2 | 2414 | ||
| AB593069.1 | 2 | 2416 | ||
| AB593070.1 | 2 | 2414 | ||
| AB593071.1 | 2 | 2414 | ||
| D82347.1 | 2 | 2424 | BAA11558.1 | |
| DN949274.1 | 1 | 91 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834477 | Chr.2:181573394 - 181728800 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map