Assay Details
Target Gene Details
Entrez Gene ID: | 223 |
Gene Name: | aldehyde dehydrogenase 9 family member A1 |
Gene Aliases: |
ALDH4, ALDH7, ALDH9, E3, TMABADH |
Location: |
Chr.1:165662212-165698663 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH9A1 | NM_000696.3 | 11 | 1769 | NP_000687.3 |
| XM_011509294.2 | 11 | 1833 | XP_011507596.1 | |
| AK293520.1 | 11 | 1673 | ||
| AK301767.1 | 11 | 1634 | ||
| AK302183.1 | 11 | 1600 | ||
| AK302191.1 | 11 | 1542 | ||
| BC151140.1 | 11 | 1757 | ||
| BC151141.1 | 11 | 1757 | ||
| U34252.1 | 11 | 1966 | AAB18827.1 | |
| U50203.1 | 11 | 1508 | AAB06721.1 | |
| X75425.1 | 10 | 1496 | CAA53176.1 |
More Information
Set Membership: |
|

Genomic Map