Assay Details
Target Gene Details
Entrez Gene ID: | 63908 |
Gene Name: | NSF attachment protein beta |
Gene Aliases: |
SNAP-BETA, SNAPB |
Location: |
Chr.20:23374519-23421519 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAPB | NM_001283018.1 | 11 | 2506 | NP_001269947.1 |
| NM_001283020.1 | 10 | 2377 | NP_001269949.1 | |
| NM_001283026.1 | 10 | 2447 | NP_001269955.1 | |
| NM_022080.2 | 11 | 2494 | NP_071363.1 | |
| NR_104266.1 | ||||
| XM_011529313.1 | 11 | 2498 | XP_011527615.1 | |
| XM_017028008.1 | 11 | 2701 | XP_016883497.1 | |
| AK022817.1 | 7 | 2020 | ||
| AK124876.1 | 11 | 2433 | ||
| AK125811.1 | 4 | 3353 | ||
| AK126340.1 | 7 | 3844 | ||
| AL834445.1 | 5 | 1910 | CAD39105.1 | |
| BC047426.1 | 10 | 2389 | AAH47426.1 | |
| BC060840.1 | 11 | 2419 | AAH60840.1 | |
| BX648330.1 |
More Information
Set Membership: |
|

Genomic Map