Assay Details
Target Gene Details
Entrez Gene ID: | 26074 |
Gene Name: | cilia and flagella associated protein 61 |
Gene Aliases: |
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4 |
Location: |
Chr.20:20052514-20360714 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP61 | NM_001167816.1 | 1 | 66 | NP_001161288.1 |
| NM_015585.3 | 2 | 172 | NP_056400.3 | |
| XM_005260688.3 | 2 | 135 | XP_005260745.1 | |
| XM_005260689.2 | 2 | 135 | XP_005260746.1 | |
| XM_005260690.3 | 2 | 135 | XP_005260747.1 | |
| XM_005260691.3 | 2 | 135 | XP_005260748.1 | |
| XM_005260692.3 | 2 | 135 | XP_005260749.1 | |
| XM_005260693.3 | 2 | 135 | XP_005260750.1 | |
| XM_005260697.1 | 2 | 135 | XP_005260754.1 | |
| XM_011529211.1 | XP_011527513.1 | |||
| XM_011529212.2 | 2 | 135 | XP_011527514.1 | |
| XM_011529213.1 | 2 | 135 | XP_011527515.1 | |
| XM_017027786.1 | 2 | 135 | XP_016883275.1 | |
| XM_017027787.1 | 2 | 135 | XP_016883276.1 | |
| XM_017027788.1 | 2 | 135 | XP_016883277.1 | |
| XM_017027789.1 | 2 | 135 | XP_016883278.1 | |
| XM_017027790.1 | 2 | 135 | XP_016883279.1 | |
| XM_017027795.1 | 2 | 135 | XP_016883284.1 | |
| AK304174.1 | 2 | 135 | ||
| AK307817.1 | ||||
| AL117439.1 | CAB55925.1 | |||
| BC028708.2 | 2 | 134 | AAH28708.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3320 | Chr.20:20051215 - 20083522 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map