Assay Details
Target Gene Details
Entrez Gene ID: | 115004 |
Gene Name: | Mab-21 domain containing 1 |
Gene Aliases: |
C6orf150, cGAS, h-cGAS |
Location: |
Chr.6:73423711-73452330 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MB21D1 | NM_138441.2 | 5 | 1523 | NP_612450.2 |
| XM_017010232.1 | XP_016865721.1 | |||
| AK097148.1 | 5 | 1479 | BAC04965.1 | |
| BC012928.1 | AAH12928.1 | |||
| BC108714.1 | 5 | 1482 | AAI08715.1 | |
| BC113606.1 | 5 | 1459 | ||
| BC113608.1 | 5 | 1459 | ||
| BC143694.1 | 5 | 1459 | ||
| KC294566.1 | 5 | 1384 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map