Assay Details
Target Gene Details
Entrez Gene ID: | 115361 |
Gene Name: | guanylate binding protein 4 |
Gene Aliases: |
Mpa2 |
Location: |
Chr.1:89181148-89198950 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP4 | NM_052941.4 | 11 | 1986 | NP_443173.2 |
| AF288814.1 | 11 | 1870 | AAL02054.1 | |
| AK131094.1 | 11 | 1899 | ||
| AK304245.1 | 7 | 898 | ||
| AK312417.1 | 11 | 1978 | ||
| AL832576.2 | 11 | 1916 | CAD89936.1 | |
| BC070055.1 | 11 | 1923 | AAH70055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830548 | Chr.1:89142066 - 89298597 on Build GRCh38 | Gain |
|
| esv2714562 | Chr.1:89133415 - 89186630 on Build GRCh38 | Deletion |
|
| nsv546796 | Chr.1:89052437 - 89256179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map