Assay Details
Target Gene Details
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLOD1 | NM_000302.3 | 13 | 1515 | NP_000293.2 |
| NM_001316320.1 | 14 | 1656 | NP_001303249.1 | |
| AK294688.1 | 13 | 1346 | ||
| AK295254.1 | ||||
| AK299150.1 | 14 | 1589 | ||
| AK312243.1 | 13 | 1448 | ||
| AK316285.1 | 14 | 1592 | ||
| BC016657.2 | 13 | 1419 | AAH16657.1 | |
| M98252.1 | 13 | 1426 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955252 | Chr.1:11963044 - 11966243 on Build GRCh38 | Deletion |
|
| nsv545427 | Chr.1:11960355 - 11966298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map