Assay Details
Target Gene Details
Entrez Gene ID: | 996 |
Gene Name: | cell division cycle 27 |
Gene Aliases: |
ANAPC3, APC3, CDC27Hs, D0S1430E, D17S978E, H-NUC, HNUC, NUC2 |
Location: |
Chr.17:47117697-47189332 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC27 | NM_001114091.2 | 19 | 3150 | NP_001107563.1 |
| NM_001256.4 | 19 | 3132 | NP_001247.3 | |
| NM_001293089.1 | 19 | 3129 | NP_001280018.1 | |
| NM_001293091.1 | 18 | 2984 | NP_001280020.1 | |
| XM_011525546.2 | 19 | 3134 | XP_011523848.1 | |
| XM_011525547.2 | 19 | 3073 | XP_011523849.1 | |
| XM_011525548.2 | 18 | 3022 | XP_011523850.1 | |
| XM_011525549.2 | 18 | 3145 | XP_011523851.1 | |
| XM_011525550.2 | 17 | 2832 | XP_011523852.1 | |
| XM_017025481.1 | 19 | 3070 | XP_016880970.1 | |
| XM_017025482.1 | 19 | 3055 | XP_016880971.1 | |
| XM_017025483.1 | 19 | 3052 | XP_016880972.1 | |
| XM_017025484.1 | 18 | 2997 | XP_016880973.1 | |
| XM_017025485.1 | 17 | 2875 | XP_016880974.1 | |
| XM_017025486.1 | 17 | 2792 | XP_016880975.1 | |
| XM_017025487.1 | 18 | 2939 | XP_016880976.1 | |
| XM_017025488.1 | 19 | 3099 | XP_016880977.1 | |
| BC011656.2 | 19 | 3014 | AAH11656.1 | |
| S78234.1 | 19 | 3092 | AAB34378.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760450 | Chr.17:47053411 - 47147856 on Build GRCh38 | Gain |
|
| dgv41n68 | Chr.17:46942994 - 47175258 on Build GRCh38 | Loss |
|
| nsv1059766 | Chr.17:47053399 - 47126102 on Build GRCh38 | Gain |
|
| esv2758692 | Chr.17:46006548 - 47199967 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map