Assay Details
Target Gene Details
Entrez Gene ID: | 10894 |
Gene Name: | lymphatic vessel endothelial hyaluronan receptor 1 |
Gene Aliases: |
CRSBP-1, HAR, LYVE-1, XLKD1 |
Location: |
Chr.11:10557866-10568818 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYVE1 | NM_006691.3 | 6 | 1146 | NP_006682.2 |
| AF118108.1 | 6 | 950 | AAD42764.1 | |
| AF127670.2 | 6 | 980 | AAD49220.2 | |
| AK075443.1 | 6 | 1060 | ||
| AK300885.1 | 4 | 593 | ||
| AK312462.1 | 6 | 1144 | ||
| AY358925.1 | 6 | 1019 | AAQ89284.1 | |
| BC026231.1 | 6 | 905 | AAH26231.1 |
Target Gene Details
Entrez Gene ID: | 100129827 |
Gene Name: | MRVI1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:10541236-10599932 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1-AS1 | NR_034093.2 | |||
| NR_034094.2 | ||||
| NR_046374.1 | ||||
| NR_046375.1 | ||||
| AI075905.1 | ||||
| BC028123.1 | AAH28123.1 | |||
| BI520309.1 | ||||
| BU569149.1 | ||||
| BU570065.1 | ||||
| DB071465.1 | ||||
| DB454301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819020 | Chr.11:10558798 - 10560604 on Build GRCh38 | Loss |
|
| nsv7675 | Chr.11:10532842 - 10565476 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map