Assay Details
Target Gene Details
Entrez Gene ID: | 205717 |
Gene Name: | upstream transcription factor family member 3 |
Gene Aliases: |
KIAA2018 |
Location: |
Chr.3:113648385-113696657 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USF3 | NM_001009899.3 | 7 | 10783 | NP_001009899.3 |
| NR_111981.1 | ||||
| XM_005247208.4 | 6 | 10711 | XP_005247265.2 | |
| XM_017005871.1 | 6 | 10678 | XP_016861360.1 | |
| XM_017005872.1 | 5 | 10595 | XP_016861361.1 | |
| BX537728.1 | 1 | 5464 | CAD97821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013506 | Chr.3:113171761 - 113962104 on Build GRCh38 | Gain |
|
| nsv819838 | Chr.3:113651010 - 113651333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map