Assay Details
Target Gene Details
Entrez Gene ID: | 282809 |
Gene Name: | POC1 centriolar protein B |
Gene Aliases: |
CORD20, PIX1, TUWD12, WDR51B |
Location: |
Chr.12:89419718-89526262 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POC1B | NM_001199777.1 | 10 | 1525 | NP_001186706.1 |
| NM_172240.2 | 11 | 1544 | NP_758440.1 | |
| NR_037659.1 | 11 | 1395 | ||
| NR_037660.1 | 9 | 1353 | ||
| AB053266.1 | 10 | 1034 | BAB69057.1 | |
| AK027829.1 | 7 | 743 | ||
| AK074772.1 | 11 | 1282 | BAC11198.1 | |
| AK123106.1 | 10 | 1522 | ||
| AK123625.1 | 7 | 742 | BAC85666.1 | |
| AK295596.1 | 11 | 1133 | ||
| AL832918.1 | 10 | 1031 | CAH10775.1 | |
| BC026080.2 | 11 | 1306 | AAH26080.1 | |
| BX648293.1 | 1 | 4145 | ||
| EF011619.1 | 11 | 1306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580371 | Chr.12:89421998 - 89431934 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map