Assay Details
Target Gene Details
Entrez Gene ID: | 8639 |
Gene Name: | amine oxidase, copper containing 3 |
Gene Aliases: |
HPAO, SSAO, VAP-1, VAP1 |
Location: |
Chr.17:42851123-42858130 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOC3 | NM_001277731.1 | NP_001264660.1 | ||
| NM_003734.3 | NP_003725.1 | |||
| NR_102422.1 | ||||
| XM_011525419.1 | XP_011523721.1 | |||
| XM_011525420.2 | XP_011523722.1 | |||
| AF067406.1 | AAC25170.1 | |||
| AK315129.1 | ||||
| BC050549.1 | AAH50549.1 | |||
| DB250074.1 | ||||
| JX020506.1 | ||||
| U39447.1 | AAC50919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map