Assay Details
Target Gene Details
Entrez Gene ID: | 100528062 |
Gene Name: | ARMCX5-GPRASP2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:102599348-102717733 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMCX5-GPRASP2 | NM_001199818.1 | NP_001186747.1 | ||
| BG392679.1 |
Target Gene Details
Entrez Gene ID: | 9737 |
Gene Name: | G protein-coupled receptor associated sorting protein 1 |
Gene Aliases: |
GASP, GASP-1, GASP1 |
Location: |
Chr.X:102651366-102659083 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPRASP1 | NM_001099410.1 | 4 | 2858 | NP_001092880.1 |
| NM_001099411.1 | 3 | 2790 | NP_001092881.1 | |
| NM_001184727.1 | 6 | 2961 | NP_001171656.1 | |
| NM_014710.4 | 5 | 2949 | NP_055525.3 | |
| XM_017029981.1 | 4 | 2838 | XP_016885470.1 | |
| XM_017029982.1 | 4 | 2873 | XP_016885471.1 | |
| AB007903.2 | 1 | 2314 | ||
| BC114552.1 | 3 | 2594 | AAI14553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526716 | Chr.X:102582805 - 102934382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map