Assay Details
Target Gene Details
Entrez Gene ID: | 219527 |
Gene Name: | leucine rich repeat containing 55 |
Gene Aliases: |
- |
Location: |
Chr.11:57181747-57191714 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC55 | NM_001005210.2 | 2 | 1080 | NP_001005210.1 |
| AK127591.1 | 2 | 1080 | BAC87047.1 | |
| BC136737.1 | 2 | 1038 | ||
| BC136739.1 | 2 | 1038 | ||
| BC150572.1 | 2 | 922 |
More Information
Set Membership: |
|

Genomic Map