Assay Details
Target Gene Details
Entrez Gene ID: | 158381 |
Gene Name: | ATPase phospholipid transporting 8B5, pseudogene |
Gene Aliases: |
FetA |
Location: |
Chr.9:35406755-35483029 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP8B5P | NR_003581.2 | |||
| NR_003582.1 | ||||
| NR_110320.1 | ||||
| AK310278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
| nsv831545 | Chr.9:35327931 - 35507322 on Build GRCh38 | Loss |
|

Genomic Map