Assay Details
Target Gene Details
Entrez Gene ID: | 2801 |
Gene Name: | golgin A2 |
Gene Aliases: |
GM130 |
Location: |
Chr.9:128255829-128276007 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA2 | NM_004486.4 | NP_004477.3 | ||
| XM_005251930.1 | XP_005251987.1 | |||
| XM_005251931.1 | XP_005251988.1 | |||
| XM_005251932.1 | XP_005251989.1 | |||
| AF248953.1 | AAF65550.1 | |||
| BC014188.2 | AAH14188.2 | |||
| BC069268.1 | AAH69268.1 | |||
| BT007248.1 | AAP35912.1 | |||
| BX647920.1 | CAI46071.1 | |||
| L06147.1 | AAA35920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972807 | Chr.9:128251140 - 128267773 on Build GRCh38 | Duplication |
|
| nsv831728 | Chr.9:128237994 - 128416514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map