Assay Details
Target Gene Details
Entrez Gene ID: | 220988 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A3 |
Gene Aliases: |
2610510D13Rik, D10S102, FBRNP, HNRPA3 |
Location: |
Chr.2:177212694-177223959 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA3 | NM_194247.2 | 11 | 4257 | NP_919223.1 |
| XM_005246380.1 | 11 | 1605 | XP_005246437.1 | |
| XM_005246381.4 | 11 | 4163 | XP_005246438.1 | |
| XM_006712365.2 | 10 | 4116 | XP_006712428.1 | |
| XM_017003587.1 | 11 | 1511 | XP_016859076.1 | |
| XM_017003588.1 | 10 | 1464 | XP_016859077.1 | |
| AL832246.1 | 5 | 770 | ||
| BX647217.1 | 10 | 1249 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834465 | Chr.2:177202566 - 177250385 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map