Assay Details
Target Gene Details
Entrez Gene ID: | 284656 |
Gene Name: | EPH receptor A10 |
Gene Aliases: |
- |
Location: |
Chr.1:37713888-37765152 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA10 | NM_001099439.1 | 15 | 2776 | NP_001092909.1 |
| XM_017001080.1 | 11 | 1518 | XP_016856569.1 | |
| XM_017001081.1 | XP_016856570.1 | |||
| AK131215.1 | 11 | 1490 | BAD18404.1 | |
| AK309089.1 | 11 | 1428 | ||
| AY726560.1 | ||||
| BC041360.1 | AAH41360.1 | |||
| BC112933.1 |
More Information
Set Membership: |
|

Genomic Map