Assay Details
Target Gene Details
Entrez Gene ID: | 51454 |
Gene Name: | GULP, engulfment adaptor PTB domain containing 1 |
Gene Aliases: |
CED-6, CED6, GULP |
Location: |
Chr.2:188291669-188595926 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954447 | Chr.2:188290974 - 188293073 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map