Assay Details
Target Gene Details
Entrez Gene ID: | 57561 |
Gene Name: | arrestin domain containing 3 |
Gene Aliases: |
TLIMP |
Location: |
Chr.5:91368724-91383359 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC3 | NM_020801.2 | 8 | 2425 | NP_065852.1 |
| AB037797.1 | 8 | 2397 | ||
| AK025028.1 | 1 | 209 | ||
| AK026917.1 | 1 | 732 | ||
| BC015928.2 | 8 | 2392 | AAH15928.1 | |
| BC053619.1 | 8 | 2397 | AAH53619.1 |
More Information
Set Membership: |
|

Genomic Map