Assay Details
Target Gene Details
Entrez Gene ID: | 8934 |
Gene Name: | RAB29, member RAS oncogene family |
Gene Aliases: |
RAB7L, RAB7L1 |
Location: |
Chr.1:205767986-205775487 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB29 | NM_001135662.1 | 6 | 1962 | NP_001129134.1 |
| NM_001135663.1 | 4 | 2177 | NP_001129135.1 | |
| NM_001135664.1 | 5 | 1809 | NP_001129136.1 | |
| NM_003929.2 | 6 | 2063 | NP_003920.1 | |
| XM_005245569.1 | 6 | 1961 | XP_005245626.1 | |
| XM_005245570.1 | 6 | 2014 | XP_005245627.1 | |
| XM_005245571.1 | 6 | 1939 | XP_005245628.1 | |
| XM_006711605.3 | 5 | 1705 | XP_006711668.1 | |
| XM_006711606.2 | 5 | 1669 | XP_006711669.1 | |
| XM_017002748.1 | 5 | 1964 | XP_016858237.1 | |
| XM_017002749.1 | 5 | 1942 | XP_016858238.1 | |
| XM_017002750.1 | 5 | 1874 | XP_016858239.1 | |
| D84488.1 | 5 | 1763 | BAA22160.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs71762778] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map