Assay Details
Target Gene Details
Entrez Gene ID: | 5067 |
Gene Name: | contactin 3 |
Gene Aliases: |
BIG-1, PANG, PCS |
Location: |
Chr.3:74262568-74614448 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTN3 | NM_020872.2 | 22 | 4124 | NP_065923.1 |
| XM_005264757.3 | 23 | 4234 | XP_005264814.1 | |
| XM_011533768.2 | 22 | 7756 | XP_011532070.1 | |
| XM_017006507.1 | 23 | 4804 | XP_016861996.1 | |
| XM_017006508.1 | 22 | 4029 | XP_016861997.1 | |
| XM_017006509.1 | 13 | 2803 | XP_016861998.1 | |
| AB040929.1 | 20 | 3773 | BAA96020.1 | |
| AK001157.1 | 1 | 104 | ||
| AL512746.1 | 2 | 1149 | CAC21671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834733 | Chr.3:74098138 - 74317596 on Build GRCh38 | Loss |
|
| nsv526832 | Chr.3:73563798 - 74732599 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map