Assay Details
Target Gene Details
Entrez Gene ID: | 23122 |
Gene Name: | cytoplasmic linker associated protein 2 |
Gene Aliases: |
- |
Location: |
Chr.3:33496245-33718356 on Build GRCh38 |
Assay Gene Location: | Within Exon 44 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460487 | Chr.3:33468269 - 33561415 on Build GRCh38 | Gain |
|
| nsv999642 | Chr.3:33457884 - 33521772 on Build GRCh38 | Gain |
|
| nsv1001132 | Chr.3:33386293 - 33565948 on Build GRCh38 | Gain |
|
| nsv1006641 | Chr.3:33478025 - 33527699 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80104935] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map