Assay Details
Target Gene Details
Entrez Gene ID: | 84688 |
Gene Name: | chromosome 9 open reading frame 24 |
Gene Aliases: |
CBE1, NYD-SP22, SMRP1, bA573M23.4 |
Location: |
Chr.9:34379019-34400833 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf24 | XM_005251616.4 | 1 | 824 | XP_005251673.1 |
Target Gene Details
Entrez Gene ID: | 203259 |
Gene Name: | family with sequence similarity 219 member A |
Gene Aliases: |
C9orf25 |
Location: |
Chr.9:34398184-34458610 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM219A | NM_001184940.1 | 6 | 1819 | NP_001171869.1 |
| NM_001184941.1 | 6 | 1816 | NP_001171870.1 | |
| NM_001184942.1 | 6 | 1783 | NP_001171871.1 | |
| NM_001184943.1 | 6 | 1765 | NP_001171872.1 | |
| NM_001184945.1 | 6 | 1732 | NP_001171874.1 | |
| NM_147202.1 | 6 | 1768 | NP_671735.1 | |
| XM_005251402.4 | 7 | 2026 | XP_005251459.1 | |
| XM_011517797.2 | 6 | 1826 | XP_011516099.1 | |
| XM_011517798.2 | 6 | 1775 | XP_011516100.1 | |
| XM_011517799.2 | 6 | 1624 | XP_011516101.1 | |
| AK022819.1 | 1 | 437 | ||
| AK091262.1 | 2 | 1159 | ||
| AK096350.1 | 6 | 1768 | ||
| AK123887.1 | 7 | 1630 | ||
| AL833397.1 | 1 | 765 | ||
| BC025348.1 | 2 | 1176 | AAH25348.1 | |
| BC041009.2 | 6 | 1732 | AAH41009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain |
|
| nsv831542 | Chr.9:34273805 - 34461547 on Build GRCh38 | Gain |
|
| nsv1015595 | Chr.9:34342674 - 34474981 on Build GRCh38 | Gain |
|
| nsv1028418 | Chr.9:34360053 - 34401766 on Build GRCh38 | Gain |
|
| nsv1016251 | Chr.9:34336344 - 34408206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map