Assay Details
Target Gene Details
Entrez Gene ID: | 51207 |
Gene Name: | dual specificity phosphatase 13 |
Gene Aliases: |
BEDP, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP |
Location: |
Chr.10:75094432-75109221 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP13 | NM_001007271.1 | NP_001007272.1 | ||
| NM_001007272.1 | NP_001007273.1 | |||
| NM_001007273.1 | NP_001007274.1 | |||
| NM_001320842.1 | NP_001307771.1 | |||
| NM_001320843.1 | NP_001307772.1 | |||
| XM_005269883.3 | XP_005269940.1 | |||
| XM_005269887.1 | XP_005269944.1 | |||
| XM_005269890.1 | XP_005269947.1 | |||
| XM_011539853.1 | XP_011538155.1 | |||
| XM_011539854.2 | XP_011538156.1 | |||
| XM_011539856.2 | XP_011538158.1 | |||
| XM_017016314.1 | XP_016871803.1 | |||
| AK057012.1 | ||||
| AK127886.1 | ||||
| AK291891.1 | ||||
| AK291897.1 | ||||
| AK300679.1 | ||||
| AY040091.1 | AAK77966.1 | |||
| AY674051.1 | AAT79356.1 | |||
| BC009778.1 | AAH09778.1 | |||
| BU580940.1 |
Target Gene Details
Entrez Gene ID: | 142891 |
Gene Name: | sterile alpha motif domain containing 8 |
Gene Aliases: |
HEL-177, SMSr |
Location: |
Chr.10:75099592-75182123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD8 | XM_011539312.2 | XP_011537614.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831913 | Chr.10:75101265 - 75261737 on Build GRCh38 | Loss |
|
| nsv516793 | Chr.10:75094806 - 75108141 on Build GRCh38 | Gain |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
| nsv7199 | Chr.10:75077582 - 75112320 on Build GRCh38 | Insertion |
|
| nsv518639 | Chr.10:75083167 - 75108141 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map