Assay Details
Target Gene Details
Entrez Gene ID: | 1577 |
Gene Name: | cytochrome P450 family 3 subfamily A member 5 |
Gene Aliases: |
CP35, CYPIIIA5, P450PCN3, PCN3 |
Location: |
Chr.7:99648189-99680026 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A5 | NM_000777.4 | 10 | 1066 | NP_000768.1 |
| NM_001291829.1 | 11 | 1566 | NP_001278758.1 | |
| NM_001291830.1 | 11 | 1326 | NP_001278759.1 | |
| NR_033807.2 | 10 | 2670 | ||
| XM_006715859.2 | 10 | 1033 | XP_006715922.1 | |
| XM_011515843.1 | 11 | 1183 | XP_011514145.1 | |
| XM_017011781.1 | 7 | 708 | XP_016867270.1 | |
| XM_017011782.1 | 5 | 505 | XP_016867271.1 | |
| AK055879.1 | 2 | 838 | ||
| AK223008.1 | 10 | 1036 | BAD96728.1 | |
| AK296205.1 | 11 | 1293 | ||
| AK313813.1 | 10 | 1033 | ||
| BC033862.1 | 10 | 1025 | AAH33862.1 | |
| BX537676.1 | 10 | 2641 | CAD97807.1 | |
| BX648856.1 | 10 | 2622 | ||
| J04813.1 | 10 | 1023 | AAA02993.1 | |
| L26985.1 | 12 | 1256 | ||
| X90579.1 | 11 | 1155 |
Target Gene Details
Entrez Gene ID: | 221785 |
Gene Name: | zinc finger and SCAN domain containing 25 |
Gene Aliases: |
ZNF498 |
Location: |
Chr.7:99616600-99738689 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN25 | XM_011515909.2 | XP_011514211.1 | ||
| XM_017011827.1 | XP_016867316.1 | |||
| BX640720.1 | CAE45839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5865 | Chr.7:99656798 - 99690110 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map