Assay Details
Target Gene Details
Entrez Gene ID: | 5468 |
Gene Name: | peroxisome proliferator activated receptor gamma |
Gene Aliases: |
CIMT1, GLM1, NR1C3, PPARG1, PPARG2, PPARgamma |
Location: |
Chr.3:12287850-12471054 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPARG | NM_005037.5 | NP_005028.4 | ||
| NM_138711.3 | 1 | 94 | NP_619725.2 | |
| NM_138712.3 | NP_619726.2 | |||
| XM_011533841.2 | 1 | 225 | XP_011532143.1 | |
| AB097931.1 | 1 | 193 | BAD20647.1 | |
| AB107271.1 | 1 | 193 | BAD34540.1 | |
| AB472042.1 | 1 | 193 | ||
| AB565476.1 | ||||
| AK290581.1 | ||||
| AL523434.3 | ||||
| BG323933.1 | ||||
| BM923992.1 | 1 | 94 | ||
| X90563.1 | CAA62152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1147269 | Chr.3:11914420 - 12871937 on Build GRCh38 | Deletion |
|
| nsv589700 | Chr.3:12286252 - 12299426 on Build GRCh38 | Loss |
|
| nsv1112379 | Chr.3:11914395 - 12871920 on Build GRCh38 | Deletion |
|
| dgv850e199 | Chr.3:11914188 - 12872013 on Build GRCh38 | Deletion |
|
| dgv763e201 | Chr.3:11909121 - 12871926 on Build GRCh38 | Deletion |
|
| nsv3702 | Chr.3:12256576 - 12301217 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map