Assay Details
Target Gene Details
Entrez Gene ID: | 54878 |
Gene Name: | dipeptidyl peptidase 8 |
Gene Aliases: |
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141 |
Location: |
Chr.15:65442463-65517713 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPP8 | NM_001320875.1 | 21 | 3588 | NP_001307804.1 |
| NM_001320876.1 | 20 | 3557 | NP_001307805.1 | |
| NM_017743.5 | 18 | 3152 | NP_060213.2 | |
| NM_130434.4 | 20 | 3452 | NP_569118.1 | |
| NM_197960.3 | 21 | 3493 | NP_932064.1 | |
| NM_197961.3 | 20 | 3435 | NP_932065.1 | |
| NR_135485.1 | 21 | 3640 | ||
| NR_135486.1 | 22 | 3623 | ||
| XM_005254505.1 | 12 | 2134 | XP_005254562.1 | |
| XM_011521730.1 | 19 | 4712 | XP_011520032.1 | |
| XM_011521731.1 | 19 | 4667 | XP_011520033.1 | |
| XM_011521732.2 | 19 | 4729 | XP_011520034.1 | |
| XM_011521733.1 | 19 | 3259 | XP_011520035.1 | |
| XM_011521734.1 | 18 | 4541 | XP_011520036.1 | |
| XM_017022373.1 | 20 | 4841 | XP_016877862.1 | |
| XM_017022374.1 | 19 | 4688 | XP_016877863.1 | |
| XM_017022375.1 | 19 | 3304 | XP_016877864.1 | |
| XM_017022376.1 | 19 | 3280 | XP_016877865.1 | |
| XM_017022377.1 | 18 | 3130 | XP_016877866.1 | |
| XM_017022378.1 | 18 | 4576 | XP_016877867.1 | |
| XM_017022379.1 | 18 | 3106 | XP_016877868.1 | |
| XM_017022381.1 | 17 | 2977 | XP_016877870.1 | |
| XM_017022382.1 | 13 | 2261 | XP_016877871.1 | |
| XM_017022383.1 | 13 | 2233 | XP_016877872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map