Assay Details
Target Gene Details
Entrez Gene ID: | 11123 |
Gene Name: | RCAN family member 3 |
Gene Aliases: |
DSCR1L2, MCIP3, RCN3, hRCN3 |
Location: |
Chr.1:24502351-24537020 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN3 | NM_001251977.1 | 4 | 773 | NP_001238906.1 |
| NM_001251978.1 | 4 | 687 | NP_001238907.1 | |
| NM_001251979.1 | 4 | 665 | NP_001238908.1 | |
| NM_001251980.1 | 3 | 522 | NP_001238909.1 | |
| NM_001251981.1 | 3 | 599 | NP_001238910.1 | |
| NM_001251982.1 | NP_001238911.1 | |||
| NM_001251983.1 | 3 | 552 | NP_001238912.1 | |
| NM_001251984.1 | 3 | 411 | NP_001238913.1 | |
| NM_001251985.1 | NP_001238914.1 | |||
| NM_013441.3 | 4 | 806 | NP_038469.1 | |
| AF176116.1 | 3 | 515 | AAF01684.1 | |
| AF176117.1 | 3 | 407 | AAF01685.1 | |
| AY906854.1 | AAW83514.1 | |||
| BC035854.1 | 4 | 806 | AAH35854.1 | |
| BX105019.1 | 2 | 233 | ||
| CT001954.1 | 3 | 546 | ||
| CX752875.1 | 4 | 674 | ||
| EF431960.1 | 2 | 345 | ||
| EF467309.1 | ||||
| EF529733.1 | 2 | 345 | ||
| HQ287726.1 | 3 | 335 | ||
| HQ317421.1 | 4 | 641 | ||
| HQ317422.1 | 3 | 387 | ||
| HQ317423.1 | 4 | 687 | ||
| HQ317426.1 | 3 | 556 | ||
| HQ317427.1 | 2 | 302 | ||
| JN203053.1 | 2 | 382 | ||
| JN228252.1 | 4 | 660 |
More Information
Set Membership: |
|

Genomic Map