Assay Details
Target Gene Details
Entrez Gene ID: | 10570 |
Gene Name: | dihydropyrimidinase like 4 |
Gene Aliases: |
CRMP3, DRP-4, ULIP4 |
Location: |
Chr.10:132184983-132205776 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYSL4 | NM_006426.2 | NP_006417.2 | ||
| XM_005252658.3 | XP_005252715.1 | |||
| XM_011539125.2 | XP_011537427.1 | |||
| XM_017015482.1 | XP_016870971.1 | |||
| XM_017015483.1 | XP_016870972.1 | |||
| AB006713.1 | BAA21886.1 | |||
| BC136329.1 | ||||
| BC136330.1 | ||||
| Y10976.1 | CAA71872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054569 | Chr.10:132009350 - 132351508 on Build GRCh38 | Gain |
|
| esv3624956 | Chr.10:131249138 - 132223150 on Build GRCh38 | Gain |
|
| esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv951885 | Chr.10:132093297 - 132212896 on Build GRCh38 | Deletion |
|
| nsv552504 | Chr.10:132180474 - 132255847 on Build GRCh38 | Loss |
|
| nsv552502 | Chr.10:132139965 - 132247418 on Build GRCh38 | Loss |
|
| nsv832030 | Chr.10:132093129 - 132236507 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114312744] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map