Assay Details
Target Gene Details
Entrez Gene ID: | 1185 |
Gene Name: | chloride voltage-gated channel 6 |
Gene Aliases: |
CLC-6 |
Location: |
Chr.1:11806096-11843144 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCN6 | NM_001256959.1 | 22 | 5173 | NP_001243888.1 |
| NM_001286.3 | 23 | 5239 | NP_001277.1 | |
| NR_046428.1 | 23 | 5223 | ||
| D28475.2 | 23 | 5124 | ||
| X83378.1 | 23 | 5098 | CAA58292.1 |
Target Gene Details
Entrez Gene ID: | 100379251 |
Gene Name: | NPPA antisense RNA 1 |
Gene Aliases: |
NPPA-AS, NPPAAS |
Location: |
Chr.1:11840319-11847616 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPPA-AS1 | NR_037806.1 | |||
| FJ706070.1 | ||||
| FJ706071.1 | ||||
| FJ706072.1 | ||||
| FJ706073.1 | ||||
| FJ706074.1 | ||||
| FJ706075.1 | ||||
| FJ706076.1 | ||||
| FJ706077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527521 | Chr.1:11840768 - 11849457 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map